Chromium ICP Standard, 1 ml = 1 mg Cr (1,000 ppm Cr)Cr(NO₃)₃ in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPCR1KN0-100N
RPCR1KN0-250N
RPCR1KN0-500N
RCPCR1KN-100EA
214.88
CAD
RCPCR1KN-100
RCPCR1KN-250
RCPCR1KN-500
Chromium ICP Standard, 1 ml = 1 mg Cr (1,000 ppm Cr)Cr(NO₃)₃ in 3% HNO₃, Ricca Chemical Company
Standards
Cationic Standards
Chromium Standards