Zinc sulfate 0.0500 M (M/20) in aqueous solution

Supplier: Ricca Chemical

R9660000-1A R9660000-500A
RC966032EA 191.18 CAD
RC966032 RC9660-16
Zinc sulfate 0.0500 M (M/20) in aqueous solution
Zinc sulphate
Formula: O₄SZn
MW: 161.45
Melting Pt: 100
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
Merck Index: 12,10293

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.05000 ± 0.00001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR