Sulfuric acid ≥75% (v/v) in aqueous solution (3 + 1)

Supplier: Ricca Chemical
Danger

R8187000-500C1 R8187000-1C1
RC818716EA 145.36 CAD
RC818716 RC818732
Sulfuric acid ≥75% (v/v) in aqueous solution (3 + 1)
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 75.0 ± 0.7 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR