Hydrochloric acid 5% (v/v) in aqueous solution (1 + 19) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

R3568000-500A R3568000-4A R3568000-1A R3568000-20F
RC3568-16EA 66.36 CAD
RC3568-16 RC3568-1 RC3568-32 RCR3568000-20F
Hydrochloric acid 5% (v/v) in aqueous solution (1 + 19) analytical reagent
Hydrochloric acid
Formula: HCl
Boiling Pt: ? 100 °C (1013 hPa)
Density: 1.02 to 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 4.96-5.04 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR