Hydrochloric acid 2% (v/v) in aqueous solution (1 + 49)
Supplier: Ricca Chemical
|
Warning
|
R3567000-4A
R3567000-500A
R3567000-1A
RC3567-1EA
181.7
CAD
RC3567-1
RC3567-16
RC3567-32
Hydrochloric acid 2% (v/v) in aqueous solution (1 + 49)
Hydrochloric acid
Formula:
HCl Boiling Pt: ∼100 °C (1013 hPa) Density: 1.02…1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 2.00 ± 0.04 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...