Hydrochloric acid 1.96% (v/v) in aqueous solution (1 + 50)
Supplier: Ricca Chemical
|
Warning
|
R3565900-500A
R3565900-1A
RC3565.9-16EA
165.9
CAD
RC3565.9-16
RC3565.9-32
Hydrochloric acid 1.96% (v/v) in aqueous solution (1 + 50)
Hydrochloric acid
Formula:
HCl Boiling Pt: ∼100 °C (1013 hPa) Density: 1.02…1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 1.96 ± 0.04 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...