Hydrochloric acid 0.0500 N (N/20) analytical reagent

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3597000-4A R3597000-1A R3597000-20F
RC35971EA 189.6 CAD
RC35971 RC359732 RC3597-5
Hydrochloric acid 0.0500 N (N/20) analytical reagent
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.05000 ± 0.00005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR