Sodium hydroxide 4% (w/v) in aqueous solution (40 g/L) analytical reagent
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda
R7247000-4A
R7247000-500A
R7247000-1A
RC72471EA
241.74
CAD
RC72471
RC724716
RC7247-32
Sodium hydroxide 4% (w/v) in aqueous solution (40 g/L) analytical reagent
Sodium hydroxide
Formula:
NaOH Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 4.00 ± 0.04 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...