Sodium hydroxide 0.0100 N (N/100) analytical reagent
Supplier: Ricca Chemical
Synonyms:
Caustic soda
R7295000-4A
R7295000-20F
R7295000-1A
R7295000-10F
R7295000-500A
RC72951EA
192.76
CAD
RC72951
RC72955
RC729532
RC7295-2.5
RCR7295000500A
Sodium hydroxide 0.0100 N (N/100) analytical reagent
Sodium hydroxide
Formula:
NaOH Melting Pt: 0.0°C Density: 1 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 0.00990-0.01010 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...