Potassium chromate 5% (w/v) in aqueous solution indicator, for argentometric titration chloride-free

Supplier: Ricca Chemical

R6000000-500A R6000000-1A R6000000-120A
RC600016EA 107.44 CAD
RC600016 RC600032 RC60004
Potassium chromate 5% (w/v) in aqueous solution indicator, for argentometric titration chloride-free
Potassium chromate
Formula: K₂CrO₄
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, yellow, odorless
Suitability for Use Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR