Potassium hydroxide 50% (w/v) in aqueous solution for Orsat gas analysis
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Potash caustic, Caustic potash
R6160000-500A
R6160000-1A
R6160000-4A
RC616016EA
162.74
CAD
RC616016
RC616032
RCH61601
Potassium hydroxide 50% (w/v) in aqueous solution for Orsat gas analysis
Potassium hydroxide
Formula:
KOH Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1814 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 50.00 ± 0.06 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...