Potassium hydroxide 1.00 N methanol analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Potash caustic, Caustic potash

R6262000-4A R6262000-1A
RC62621EA 587.76 CAD
RC62621 RC626232
Potassium hydroxide 1.00 N methanol analytical reagent
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 84) 1.000 ± 0.005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR