Potassium hydroxide 0.500 N (N/2) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Potash caustic, Caustic potash
R6230000-4A
R6230000-1A
R6230000-20F
RC62301EA
339.7
CAD
RC62301
RC623032
RC62305
Potassium hydroxide 0.500 N (N/2) in aqueous solution
Potassium hydroxide
Formula:
KOH MW: 56.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1814 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.5000 ± 0.0002 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...