Potassium hydroxide 0.100 N (N/10) in methanol

Supplier: Ricca Chemical
Danger

R6260000-4A R6260000-1A
RC62601EA 450.3 CAD
RC62601 RC626032
Potassium hydroxide 0.100 N (N/10) in methanol
Potassium hydroxide
Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 84) 0.1000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR