Potassium hydroxide 0.100 N (N/10)

Supplier: Ricca Chemical
Warning

Synonyms: Potash caustic, Caustic potash

R6200000-4A R6200000-1A R6200000-20F R6200000-500A
RC62001EA 323.9 CAD
RC62001 RC620032 RC62005 RCR6200000500A
Potassium hydroxide 0.100 N (N/10)
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 0.0°C
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.0999-0.1001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR