Acetic acid 2.00 N analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

R0151000-4A R0151000-500A
RC151-1EA 265.44 CAD
RC151-1 RC151-16
Acetic acid 2.00 N analytical reagent
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, strong vinegar odor
Assay at 20 °C (traceable to NIST SRM 84) 2.000 ± 0.002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR