Aluminum Standards for AA Spectroscopy, Ricca Chemical Company
Supplier: Ricca Chemical
R0605000-500A R0605000-120A RAAL1KH0-100N RAAL1KN0-100N
RC60516EA 421.86 CAD
RC60516 RC6054 RCAAL1KH-100 RCAAL1KN-100 RCS0150000-1N RCS0150000-5N
Aluminum Standards for AA Spectroscopy, Ricca Chemical Company
Standards Cationic Standards Aluminum Standards
Caution: 10,000 ppm Al = 10,000 mg/l Al = 10 mg/ml Al= 10,000 µg/ml Al