EDTA disodium salt 0.0500 M (M/20) in aqueous solution

Supplier: Ricca Chemical

R2705000-4A R2705000-1A R2705000-20F
RC27051EA 282.82 CAD
RC27051 RC270532 RC27055
EDTA disodium salt 0.0500 M (M/20) in aqueous solution
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.05000 ± 0.00001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR