Hydrochloric acid 6.00 N analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

R3750000-4A R3750000-1A R3750000-10F R3750000-20F R3750000-500A
RC37501EA 336.54 CAD
RC37501 RC375032 RC3750-2.5 RC3750-5 RC3750-16
Hydrochloric acid 6.00 N analytical reagent
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow, Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 6.000 ± 0.005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR