EDTA disodium salt 0.0575 M in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
R2710000-4A
R2710000-1A
R2710000-20F
R2710000-10F
RC27101EA
295.46
CAD
RC27101
RC271032
RC27105
RC2710-2.5
EDTA disodium salt 0.0575 M in aqueous solution
EDTA disodium salt
Formula:
C₁₀H₁₄N₂Na₂O₈ MW: 336.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070672 CAS Number: 139-33-3 EINECS: 205-358-3 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 0.05750 ± 0.00005 M at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...