EDTA disodium salt 0.0200 M (M/50) in aqueous solution analytical reagent

Supplier: RICCA CHEMICAL CO CA

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

R2702000-4A R2702000-1A R2702000-20F
CARC2702-1EA 167.27 CAD
CARC2702-1 CARC2702-32 CARC2702-5
EDTA disodium salt 0.0200 M (M/50) in aqueous solution analytical reagent
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.02000 ± 0.00001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR