Potassium thiocyanate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
R6580000-4A
R6580000-1A
RC65801EA
320.74
CAD
RC65801
RC658032
Potassium thiocyanate 0.100 N (N/10) in aqueous solution
Potassium thiocyanate
Formula:
KSCN Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.1000 ± 0.0001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...