Oxalic acid 10% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R5470000-500A R5470000-1A
RC547016EA 115.34 CAD
RC547016 RC5470-32
Oxalic acid 10% (w/v) in aqueous solution analytical reagent
Oxalic acid
Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 10.0 ± 0.5 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR