Hydrochloric acid 1% (v/v) in aqueous solution (1 + 99)

Supplier: Ricca Chemical
Danger

R3560000-500A R3560000-1A R3560000-20F R3560000-4A
RC356016EA 61.62 CAD
RC356016 RC356032 RC35605 RCR3560000-4A
Hydrochloric acid 1% (v/v) in aqueous solution (1 + 99)
Hydrochloric acid
Formula: HCl
Boiling Pt: ? 100 °C (1013 hPa)
Density: 1.02 to 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.96-1.04 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR