Hydrochloric acid 25% (v/v) in aqueous solution (1 + 3) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

R3577000-4A R3577000-500A R3577000-1A
RC3577-1EA 349.18 CAD
RC3577-1 RC357716 RC357732
Hydrochloric acid 25% (v/v) in aqueous solution (1 + 3) analytical reagent
Hydrochloric acid
Formula: HCl
MW: 36.46
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 25.0 ± 0.1 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR