Cryule® Vial Freezer Box
Supplier: DWK Life Sciences (KIMBLE)
651490 651492
16300-192CS 423.77 CAD
16300-192 16300-193
Cryule® Vial Freezer Box
The WHEATON Cryule® Freezer box stores 1 and 2 mL cryogenic ampules and vials
Box dimensions are the following: 1 mL Cryule Freezer Box - 133 x 133 x 32 mm; 2 mL Cryule Freezer Box - 133 x 133 x 44 mm.
Made from polyethylene coated chipboard, it is designed for use in the vapor phase of liquid nitrogen. The box is designed to hold 81 freestanding or round bottom vials or ampules.