CryoELITE® Cryogenic Vials, Round Bottom, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W985900 W985901 W985910 W985911 W985912 W985913 W985914 W985850 W985851 W985860 W985861
89125-446CS 1277.13 CAD
89125-446 89125-448 89125-454 89125-456 89125-458 89125-460 89125-462 89125-486 89125-488 89125-494 89125-496
CryoELITE® Cryogenic Vials, Round Bottom, Wheaton
Vials
Vials are available with either a patented 2' angle plug seal for external thread vials or with an internal thread.
  • Made from low binding, cryogenic grade virgin polypropylene
The availability of colored caps allows for the color coding of projects along with WHEATON colored freezer and storage boxes.
Both external and internal CryoELITE offer unique stop-rims to provide additional security in knowing the cap is securely closed. WHEATON CryoELITE® Cryogenic Round Bottom Vials have an exacting bottom that allows for up to 17000 MAX RCF (×g). The patented unrivaled cap seal exceeds DOT and IATA regulations, ensuring the ultimate level of protection for samples during transportation and demanding free-thaw handling.
Certifications: Each lot is certified RNase/DNase and endotoxin free and non-pyrogenic, providing assurance for sample integrity.