PolarSafe™ Stainless Steel Specialty Freezer Racks, Argos Technologies
Supplier: Antyila Scientific
04400-09 04398-34 04399-94 04400-07 04400-47 04400-48 04400-50 04398-36 04400-96 04401-00 04401-02 04398-35 04399-99 04401-94 04401-97 04401-98 04401-99 04402-02 04402-03 04402-04 04402-09 04402-10 04402-11 04402-12 04402-13 04402-14 04402-15 04402-16 04402-17 04402-33 04402-34 04402-35 04402-40 04402-41 04402-42 04402-43 04402-45 04402-46 04402-47 04402-60 04402-62 04402-64 04402-71 04402-90 04402-91 04402-94 04402-96 04402-97 04402-98 04403-11 04403-13 04403-15 04403-16 04403-19
71001-392EA 817.24 CAD
71001-392 71001-394 71001-396 71001-398 71001-416 71001-418 71001-420 71001-422 71001-432 71001-434 71001-436 71001-438 89128-172 89128-174 89220-540 89220-542 89220-544 89220-546 89220-548 89220-550 89220-552 89220-554 89220-556 89220-558 89220-560 89220-562 89220-564 89220-566 89220-568 89220-570 89220-572 89220-574 89220-596 89220-598 89220-600 89220-602 89220-604 89220-608 89220-610 89220-612 89220-614 89220-616 89220-618 89220-620 89220-622 89220-624 89220-626 89220-628 89220-630 89220-632 89220-634 89220-636 89220-638 89220-640 89220-642 89220-644 89220-646 89220-648 89220-650 89220-652 89220-654 89220-656 89220-658 89220-660 89220-662 89220-664 89220-666 89220-668 89220-670 89220-672 89220-674 89220-676 89220-678 89220-680 89220-682 89220-684 89220-686 89220-688 89220-690
PolarSafe™ Stainless Steel Specialty Freezer Racks, Argos Technologies
Racks Cryogenic Box Racks
Argos PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel
Argos PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.
Handles at both ends accommodate freezers with right or left handed doors.
Ordering information: Additional freezer racks available. Contact your Avantor sales representative for details.