Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
121-0020 121-0045 122-0020 122-0045 124-0045 524-0020
28199-030CS 1494.5 CAD
28199-030 28199-032 28199-034 28199-036 28199-048 73520-980
Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Filters Bottle Top Filters
Complete filter units.
  • Choose from CN, aPES, PES, or SFCA membranes
  • Use 0.2 µm pore size for sterile filtration and 0.45 µm pore size for clarification and particle removal
Note: For research use only. Not for in vitro diagnosis or parenterals.
Presterilized, individually packaged one-piece vacuum filter units with 115 ml capacity are ideal for easy, economical filtration of 50 to 115 ml of aqueous fluids. Upper and lower chambers with graduations do not separate; simply remove vacuum hose after filtration and pour filtered media out. CatalogMore Product Information