Dropping Bottle, LDPE, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W242826 W242832 W242823 W242822-A W242825-A W242834-A W242837 W242833 W242836-A W242832-A W242835-A W242831-A W242834 W242836 W242826-A W242831 W242823-A W242821 W242833-A W242825 W242824 W242824-A W242835 W242822 W242821-A W242827
16300-362CS 458.45 CAD
16300-362 16300-384 16300-340 16300-342 16300-386 16300-388 16300-344 16300-368 16300-382 16300-360 16300-338 16300-352 16300-374 16300-376 16300-354 16300-332 16300-356 16300-378 16300-334 16300-336 16300-358 16300-390 16300-370 16300-392 16300-350 16300-372
Dropping Bottle, LDPE, WHEATON®, DWK Life Sciences
Bottles Dropping Bottles
WHEATON Dropping Bottles are made of natural or white LDPE (Low Density Polyethylene)
White LDPE bottles help protect UV light sensitive samples. Dropping bottles are packaged in cases of 100, 1000 or bulk.