Vacule® Lyophilization Ampules, Pre-scored, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
Vacule®
651502
26675-887CS 373.84 CAD
26675-887
Vacule® Lyophilization Ampules, Pre-scored, WHEATON®, DWK Life Sciences
Ampoules
Vacules are made from low extractable borosilicate glass.
  • Pre-scored to eliminate the need for filing
The ampule is ideal for small volume lyophilization samples
Ext.Ø: 10 mm at top
Certifications: USP Type 1, ASTM E 438 Type 1, Class A