Cryule® Cryogenic Ampule, Pre-scored, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
651483 651486
16300-190CS 283.42 CAD
16300-190 16300-191
Cryule® Cryogenic Ampule, Pre-scored, WHEATON®, DWK Life Sciences
Ampoules
Cryules are made from low extractable borosilicate glass.
  • Pre-scored to eliminate the need for filing
  • Disposable Ampule Snapper protects the user against glass splinters; prevents cross contamination between samples
For use in biological sample preservation in glass. Cryules can be used in the vapor phase of liquid nitrogen and are specially designed for storage at low temperatures as well as rapid thawing.
Ext.Ø: 5 mm at top
Certifications: USP Type 1, ASTM E 438 Type I, Class A