WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
242401 242402 242403 242404 242405 242406 242407 242408 242410 242411 242412 242413 242415 242416 242420 242421 242422 242423 242424 242425 242426 242427 242430 242431 242432 242433 242434 242436 242437 242438 242440 W242401-01-A W242410-A W242420-A 242414 242401-01 242411-01 242431-01 W242402-A W242404-A W242406-A W242411-01-A W242411-A W242412-A W242413-A W242414-A W242415-A W242416-A W242418-A W242431-01-A W242431-A W242432-A W242433-A W242434-A W242436-A W242437-A W242438-A W242440-A W242401-A W242403-A W242405-A W242407-A W242408-A W242417-A 242421-01 W242421-01-A W242421-A W242422-A W242423-A W242424-A W242425-A W242426-A W242427-A W242428-A W242430-A
16300-062CS 66.45 CAD
16300-062 16300-063 16300-064 16300-065 16300-066 16300-067 16300-068 16300-069 16300-071 16300-072 16300-073 16300-074 16300-076 16300-077 16300-078 16300-081 16300-082 16300-084 16300-085 16300-086 16300-087 16300-088 16300-089 16300-092 16300-093 16300-094 16300-095 16300-096 16300-098 16300-099 16300-100 16300-102 80086-304 89048-954 89048-956 89220-202 89220-206 89220-208 89220-210 89220-212 89220-216 89220-220 89220-224 89220-226 89220-230 89220-234 89220-238 89220-242 89220-246 89220-250 89220-254 89220-256 89220-260 89220-264 89220-268 89220-276 89220-280 89220-284 89220-288 89500-054 89500-056 89500-058 89500-060 89500-062 89500-064 89500-066 89500-068 89500-070 89500-072 89500-074 89500-076 89500-078 89500-080 89500-082 89500-084 89500-086
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Closures Bottle Closures
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).