Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
211621 211622 211623 211625 211630 211631 211632 211633 211635 211624 211620 211634
16347-024CS 707 CAD
16347-024 16347-026 16347-028 16347-032 16347-050 16347-052 16347-054 16347-070 16347-074 16364-980 16347-022 16347-072
Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
Bottles Dropping Bottles
WHEATON Dropping Bottles are made of LDPE (Low Density Polyethyleye)
The extended controlled dropper tip dispneses one drop at a time, with an average drop size of 44 µl ± 6 µl. Radiation grade polypropylene screw caps and tips packaged separately. Dropping bottles, natural tips and white caps are packaged separately.
Dropping bottles with tips can be used to dispense a stream of liquid or a non-controlled drop. Dropping bottles, tips and caps are packaged separately and bottles are autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable