HyClone™ AdvanceSTEM™ Cryopreservation Media, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30894.01 SH30894.01
CA95053-252EA 217.08 CAD
CA95053-252 95053-252
HyClone™ AdvanceSTEM™ Cryopreservation Media, Cytiva
Cell Freezing Media
HyClone AdvanceSTEM Cryopreservation Medium is formulated to support the cryopreservation of a wide range of cell types
Formulated to support the cryopreservation of a variety of cell types, including Human Mesenchymal Stem Cells, multipotent unrestricted stem cells of Umbilical Cord Blood and Human Amniotic Epithelial Stem Cells.
Caution: Storage < -10° C