Wide Mouth Blake Packer, HDPE, Natural, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
209683 209685 209686
16153-527CS 619.96 CAD
16153-527 16153-531 16153-533
Wide Mouth Blake Packer, HDPE, Natural, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
Wheaton wide mouth blake packers are manufactured from high density polyethylene.
High Density Polyethylene (HDPE) Characteristics: Flexible but more rigid than LDPE. Natural color is milky white, semi-translucent depending on density. White HDPE helps protect light sensitive samples. Good impact strength and stress crack resistance. Good chemical resistance. Good vapor barrier but poor gas barrier. Can be sterilized using ethylene oxide (EtO) or gamma radiation.
The wide mouth is ideal for solid samples. Pre-attached white polypropylene screw cap with foamed polyethylene liner provides cleanliness. Blake design maximizes storage space.