PYREX® Reagent Bottle, Narrow Mouth, with Stopper, Corning
Supplier: Corning
1500-250 1500-2L 1500-1L 1500-500
89090-790PK 95.88 CAD
89090-790 89090-860 89090-872 89090-878
PYREX® Reagent Bottle, Narrow Mouth, with Stopper, Corning
Bottles Glass Bottles
PYREX® borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers. Replacement stoppers are available.