PYREX® Delong Shaker Erlenmeyer Flask with Extra Deep Baffles, Corning
Supplier: Corning
4446-50 4446-125 4446-250 4446-500 4446-1L 4446-2L 4446-3L 4446-4L
22877-262CS 846.85 CAD
22877-262 22877-264 22877-266 22877-268 22877-270 22877-272 22877-274 22877-276 22877-278
PYREX® Delong Shaker Erlenmeyer Flask with Extra Deep Baffles, Corning
Flasks Erlenmeyer Flasks
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These PYREX® Delong style shaker flasks have three extra deep baffles designed to provide greater agitation of solutions to improve oxygen or gas transfer when used with rotary or reciprocating shakers. The long neck reduces splashing and is designed for two-position polypropylene closures. Their uniform wall thickness provides the proper balance between mechanical strength andMore Product Information