Sampule® Vials, Glass, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
986491 986492
66021-293CS 837.8 CAD
66021-293 66021-307
Sampule® Vials, Glass, Wheaton, DWK Life Sciences
Vials
Wheaton Sampule vials are designed for liquid scintillation counting.
  • Background counts are consistent and low while ultraviolet transmission is high
Vials are manufactured from Wheaton 180 low potassium borosilicate glass that conforms to ASTM E438 Type I, Class A requirements.
Packaging: Caps are packaged separately.