Barnstead™ Pacific™ RO Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50132385 50132386 50132388
89230-436EA 7461.06 CAD
89230-436 89230-438 89230-440 89230-442 89230-444
Barnstead™ Pacific™ RO Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead Pacific RO water purification systems provides up to 240L/day of Type 3 (Reverse Osmosis) water from tap water
The Pacific RO systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. The 30L and 60L reservoirs may also be mounted on the wall, on the bench, or under the bench.
The Pacific RO systems are ideal for producing type 3 (RO) for the rinsing of glassware, supplying lab equipment such as autoclaves, chambers, or water baths, or the preparation and diluting of buffers, reagents and media.
All Pacific RO systems include: quick discconnects to make changing consumables easy, and all core consumables and tubing. Pacific RO systems are offered in two output volumes, 7L/hr and 20L/hr. External reservoirs are required and are offered in three different sizes, 30L, 60L or 100L.
Certifications: CSA, CE.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz) and all necessary tubing to connect to water inlet supply. All Pacific RO systems require the purchase of an external reservoir. Feed water must be tap water.