Extraction Condensers, Allihn, Ace Glass
Supplier: Ace Glass
6740-06 6740-26 6740-02 6740-10 6740-08 6740-04
89055-984EA 1147.2 CAD
89055-984 89055-982 89055-980 89055-978 89055-976 89055-974
Extraction Condensers, Allihn, Ace Glass
Condensers
Bulb-type Allihn condensers are for use with regular extraction apparatus
Improved design permits greater condensing capacity. Approximately one bulb per 50mm of jacket length. Standard taper extractor joint at bottom, open tube at top. Hose barbs at top and bottom for inlet/outlet. Fabricated from borosilicate glass.