SP Bel-Art FRIO-Temp® Certified Precision Liquid-In-Glass Verification Thermometer, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60200-0500 B60200-0700 B60200-0800 B60200-0900
89217-984EA 597.52 CAD
89217-984 89217-986 89217-990 89217-994 89217-998
SP Bel-Art FRIO-Temp® Certified Precision Liquid-In-Glass Verification Thermometer, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST, DKD/PTB and most standards through ILAC.
  • White back glass
  • Organic thermometer liquid
  • Highly accurate readings
  • PFA safety coating
  • Individually packaged
Each item features PFA safety coating, a detailed Calibration Report stating corrections to one-tenth of the smallest division and individual serial number. Each item is tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB. Individually packaged. Glass Color: White.
Certifications: NIST, DKD/PTB