SP Bel-Art FRIO-Temp® High Precision Verification Thermometer, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61001-0100 B61001-0200 B61001-0300 B61001-0400 B61001-0500
89217-856EA 220.69 CAD
89217-856 89217-858 89217-860 89217-862 89217-864
SP Bel-Art FRIO-Temp® High Precision Verification Thermometer, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert thermometer into the bottle and place near the samples for best results
  • Feature permanently fused markings and white lead free glass
Each item features PFA safety coating, a multi-language (English, French, German, Italian, Portuguese, Spanish) Statement of Accuracy and Traceability and individual serial number. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications: NIST.