Cryo.s™ Storage Box, Rack and Lid, Greiner Bio-One
Supplier: Greiner Bio-One
802501 802202 802203 802204 802206 802225
82051-490EA 32.71 CAD
82051-490 89176-318 89218-648 89218-650 89218-652 89218-654
Cryo.s™ Storage Box, Rack and Lid, Greiner Bio-One
Boxes Cryoboxes
Greiner Bio-One offers the Cryo.s™ Storage Box is a helpful accessory for storing Cryo.s™ sample tubes at low and ultra-low temperatures.
  • Polypropylene construction
  • Shock-resistant
  • Resilient to extremely low temperatures
The box material polypropylene is very temperature and shock-resistant, thus allowing storage temperatures as low as -196 °C.