DuPont™ Tychem® 10000 Encapsulated Level A Suits with EX Extra Wide Visor
Supplier: DuPont
TK555TLYMD000100 TK555TLYLG000100 TK555TLYXL000100 TK555TLY2X000100 TK555TLY3X000100 TK555TLY4X000100
CA10010-352EA 9713.83 CAD
CA10010-352 CA10010-354 CA10010-356 CA10010-358 CA10010-360 CA10010-362
DuPont™ Tychem® 10000 Encapsulated Level A Suits with EX Extra Wide Visor
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Encapsulated Level A suit
  • Expanded back
  • Rear entry
  • Extra-wide EX visor
  • Attached dual-layered gloves
  • Multi-layer laminate interior gloves
  • Butyl exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Double taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing. Expanded faceshield provides wide 220 degree viewing angle. EX faceshield is triple-layered (laminate composed of 40 mil PVC/5 mil Teflon® /20 mil PVC). Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment. Expanded back to accommodate self-contained air breathing apparatus (SCBA). Coveralls also feature a vapor tight zipper that is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Butyl B 161 gloves provide mechanical and physical durability to help protect inner glove. Gloves are attached with a vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system provides additional support and improved fit. Additional barrier material is sewn in at knees and over taped for added abrasion resistance when kneeling or crawling. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional.