DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Supplier: DuPont
Tychem®
SL122TWH5X0006BN SL122TWH2X00 SL122TWHMD0006BN SL122TWH3X00 SL122TWHXL0006BN SL122TWH4X00 SL122TWH3X0006BN SL122TWH5X000600 SL122TWHMD00 SL122TWH6X000600 SL122TWHXL00 SL122TWH3X00 SL122TWH4X00 SL122TWHLG0006BN SL122TWHLG00 SL122TWHLG00 SL122TWH2X00 SL122TWH2X0006BN SL122TWH4X0006BN SL122TWHMD00 SL122TWHXL00
CA13451-044CS 2569.07 CAD
13451-044 CA13451-044 13451-034 CA13451-042 CA13451-036 13451-036 CA13451-034 13451-038 10016-788 CA13451-038 10016-780 10016-792 10016-782 10016-784 10016-786 13451-040 CA13451-040 10016-480 13451-042 10016-790 10016-482
DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Coveralls feature an attached hood with elastic around face opening and elastic openings at wrists for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Integrated socks are composed of the same garment material.
Certifications: Thickness meets ASTM D1777.