Corning® DeckWorks Reload System Starter Kits, Corning
Supplier: Corning
4130 4131 4132 4133 4134
89136-144CS 78.29 CAD
89136-144 89136-146 89136-148 89136-150 89136-152
Corning® DeckWorks Reload System Starter Kits, Corning
Pipette Tips
Corning® DeckWorks™ pipette tip system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials.
  • Certified free of RNase, DNase, pyrogens and PCR inhibitors
Corning DeckWorks starter kits contain one DeckWorks reload system and empty hinged racks (10 empty racks accompany 10, 200 and 300 µl volume tips; eight empty racks acommpany 1000 µl volume tips). DeckWorks reload systems are convenient and economical - reload decks are packaged in transparent trays for easy tip identification, require noMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing