Corning® DeckWorks Reload Systems, Corning
Supplier: Corning
4125 4126 4127 4128 4129 4153 4154 4155
89136-135CS 809.58 CAD
89136-135 89136-137 89136-139 89136-141 89136-143 89136-187 89136-189 89136-191
Corning® DeckWorks Reload Systems, Corning
Pipette Tips
Corning® DeckWorks™ pipette tip system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials.
  • Certified free of RNase, DNase, pyrogens and PCR inhibitors
Corning DeckWorks reload systems are available in 10, 200, 300 and 1000 µl volumes and allow for convenient and economical reloading of durable Corning DeckWorks hinged racks. DeckWorks reload decks are packaged in transparent trays for easy tip identification, require no transfer devices, save space and minimize packaging waste. Although theMore Product Information