DuPont™ Tychem® 2000 Sleeved Aprons
Supplier: DuPont
QC278BYL00001200 QC275BYLSM002500 QC275BYLMD002500 QC275BYLLG002500 QC275BYLXL002500 QC275BYL2X002500 QC275BYL3X002500 QC275BYL4X002500
CA10010-264CS 670.99 CAD
CA10010-264 CA13452-666 CA13452-668 CA13452-670 CA13452-672 CA13452-674 CA13452-676 CA13452-678
DuPont™ Tychem® 2000 Sleeved Aprons
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Neck to knee coverage
  • Elastic wrists
  • Neck loop with snaps and waist ties
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Knee-length apron gownMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Sleeve or leg length Long
Garment application Food,Industrial,Chemical protection,Pharmaceutical