Admittance and Security Caution Signs, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
C135AB C135P C135PB C135R C135RB C41P C41PB C41R C41RB C475AB C475PB C475RB C597AB C597PB C597RB C616AB C616PB C616RB C665AC C665AD C665RC C665RD CBA9P CBA9R M96R
94004-714EA 94.42 CAD
94004-714 94004-716 94004-718 94004-720 94004-722 94004-724 94005-254 94005-256 94005-258 94005-260 94005-830 94005-832 94005-834 94005-836 94005-838 94005-840 94005-842 94005-844 94005-846 94006-060 94006-062 94006-064 94006-114 94006-116 94006-118 94006-192 94006-194 94006-196 94006-542 94006-544 94006-546 94006-736 94006-738 94006-740 94007-000 94007-002 94007-004 94007-006 94007-008 94007-010 94007-012 94007-138 94007-140 94007-142 94007-434 94007-436 94007-438 94007-440 94007-666 94007-668 94017-484 94017-486
Admittance and Security Caution Signs, National Marker
Signs
Alert visitors and guest of security policies and direct them towards appropriate entrances
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Each sign header reads "Caution".